missing translation for 'onlineSavingsMsg'
Få mere at vide

3,3'-Dichlorobenzidine-d6, TRC

Artikelnummer. 30376479
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376479 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30376479 Leverandør Toronto Research Chemicals Leverandørnr. TRCD43191325MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 3,3'-DichlorobenzidineD6(ringD6)
CAS 93951-91-8
Renhedsgrad noter HPLC
Molekylær formel C12 2H6 H4 Cl2 N2
Mængde 25 mg
Synonym [1,1'-Biphenyl-2,2',3,3',6,6'-d6]-4,4'-diamine, 5,5'-dichloro- (9CI), 3,3'-Dichlorobenzidine D6 (ring D6), 3,3'-Dichlorobenzidine-d6
InChI formel InChI=1S/C12H10Cl2N2/c13-9-5-7(1-3-11(9)15)8-2-4-12(16)10(14)6-8/h1-6H,15-16H2/i1D,2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c(c([2H])c(Cl)c1N)c2c([2H])c([2H])c(N)c(Cl)c2[2H]
IUPAC navn 4-(4-amino-3-chloro-2,5,6-trideuteriophenyl)-2-chloro-3,5,6-trideuterioaniline
Molekylvægt (g/mol) 259.16
Formel vægt 258.0598
Procent renhed >95
Vis mere Vis mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.