Få mere at vide
3,3'-Dichlorobenzidine-d6 (rings-d6), CDN
Artikelnummer. 30315012
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315012 0.01 g
30310048 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315012 Leverandør CDN Leverandørnr. CDND27030.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,3'-DichlorobenzidineD6(ringD6)
CAS 93951-91-8
Molekylær formel C12 2H6 H4 Cl2 N2
Mængde 0.01 g
Synonym [1,1'-Biphenyl-2,2',3,3',6,6'-d6]-4,4'-diamine, 5,5'-dichloro- (9 CI), 3,3'-Dichlorobenzidine D6 (ring D6), 3,3'-Dichlorobenzidine-d6
InChI formel InChI=1 S/C12H10Cl2N2/c13-9-5-7(1-3-11(9)15)8-2-4-12(16)10(14)6-8/h1-6 H,15-16H2/i1D,2 D,3 D,4 D,5 D,6 D
SMIL [2 H]c1c([2 H])c(c([2 H])c(Cl)c1N)c2c([2 H])c([2 H])c(N)c(Cl)c2[2 H]
IUPAC navn 4-(4-amino-3-chloro-2,5,6-trideuteriophenyl)-2-chloro-3,5,6-trideuterioaniline
Molekylvægt (g/mol) 259.16
Formel vægt 258.0598 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity