Få mere at vide
3,3-Di(methyl-d3)-butanoyl-4,4,4-d3 Chloride, TRC
Artikelnummer. 30375544
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30375544 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375544 Leverandør Toronto Research Chemicals Leverandørnr. TRCD456877100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,3-Di(methyl-d3)-butanoyl-4,4,4-d3 Chloride
CAS 1399295-02-3
Molekylær formel C6 D9 H2 Cl O
Mængde 100 mg
Synonym tert-Butylacetyl Chloride-d9, 3,3-Dimethylbutanoyl Chloride-d9
InChI formel InChI=1S/C6H11ClO/c1-6(2,3)4-5(7)8/h4H2,1-3H3/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])C(CC(=O)Cl)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 4,4,4-trideuterio-3,3-bis(trideuteriomethyl)butanoyl chloride
Molekylvægt (g/mol) 143.659
Formel vægt 143.106