missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Chlorophenylsulfonylamino)cyclohexanecarboxylic acid, 96%, Thermo Scientific™

Catalog Number 15438535
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
690646-00-5
C13H16ClNO4S
MFCD20040116
1-3-chlorophenylsulfonamido cyclohexanecarboxylic acid, 1-3-chlorophenyl sulfonamido cyclohexane-1-carboxylic acid, 1-3-chlorophenyl sulfonylamino cyclohexane-1-carboxylic acid, 1-3-chlorobenzenesulfonamido cyclohexane-1-carboxylic acid
This item is not returnable. View return policy
690646-00-5
C13H16ClNO4S
MFCD20040116
1-3-chlorophenylsulfonamido cyclohexanecarboxylic acid, 1-3-chlorophenyl sulfonamido cyclohexane-1-carboxylic acid, 1-3-chlorophenyl sulfonylamino cyclohexane-1-carboxylic acid, 1-3-chlorobenzenesulfonamido cyclohexane-1-carboxylic acid

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 199°C to 200°C
Quantity 250 mg
Formula Weight 317.79
Percent Purity 96%
Chemical Name or Material 3-(3-Chlorophenylsulfonylamino)cyclohexanecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'