missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(3-Chlorophenyl)-1-methyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63837.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15470749

  • 1774.60 DKK / 1g


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27665238
3-(3-Chlorophenyl)-1-methyl-2-thioxo-4-imidazolidinone

Specifications

3-(3-Chlorophenyl)-1-methyl-2-thiohydantoin
MFCD27665238
240.71
95%
1g
3-(3-Chlorophenyl)-1-methyl-2-thioxo-4-imidazolidinone
Solid
Videos
SDS