missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(3-Bromophenyl)-2-thioxo-4-thiazolidinone, 95%

Brand:  Alfa Aesar™ H63634.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15470429

  • 2296.54 DKK / 1g


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD00175734
3-(3-Bromophenyl)rhodanine

Specifications

3-(3-Bromophenyl)-2-thioxo-4-thiazolidinone
MFCD00175734
288.18
95%
1g
3-(3-Bromophenyl)rhodanine
Solid
Videos
SDS