Learn More
Alfa Aesar™ 3-(3-Bromophenyl)-1-phenyl-2-thiohydantoin, 95%
Brand:  Alfa Aesar™ H64677.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15401989
  • 5462.57 DKK / 5g
Estimated Shipment date
from Supplier 17-10-2018
Add to basket
Specifications
Chemical Identifiers
MFCD27665302
3-(3-Bromophenyl)-1-phenyl-2-thioxo-4-imidazolidinone
Specifications
3-(3-Bromophenyl)-1-phenyl-2-thiohydantoin
95%
3-(3-Bromophenyl)-1-phenyl-2-thioxo-4-imidazolidinone
Solid
5g
MFCD27665302
347.23