Learn More
3,3',5-Triiodo-L-thyronine, TRC
Product Code. 30009719
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009719 500 mg 500mg
30004472 250 mg 250mg
30010667 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30009719 Supplier Toronto Research Chemicals Supplier No. TRCT795380500MG
 Restricted Item
Forventet forsendelse: 03-12-2026
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6893-02-3
Molecular Formula C15 H12 I3 N O4
Molecular Weight (g/mol) 650.97
Synonym Liothyronine, L-Tyrosine, O-(4-hydroxy-3-iodophenyl)-3,5-diiodo-, Alanine, 3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]-, L- (8CI), Thyronine, 3,3',5-triiodo-, L- (6CI), O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine, 3,3',5-Triiodo-L-thyronine, 3,3',5-Triiodothyronine, 3,5,3'-Triiodothyronine, 4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodophenylalanine, Cyronine, L-3,3',5-Triiodothyronine, L-Liothyronine, L-T3, L-Triiodothyronine, Liothyronin, NSC 80203, T3, T3 (Hormone), T3 (amino acid), Tresitope, Triiodo-L-thyronine, Triiodothyronine, Tri-iodothyronine
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoic acid
SMILES N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C(=O)O
Specifications
InChI Formula InChI=1S/C15H12I3NO4/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22/h1-4,6,12,20H,5,19H2,(H,21,22)/t12-/m0/s1
Quantity 500 mg
Formula Weight 650.79
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material 3,3',5-Triiodo-L-thyronine (T3)
Grade HPLC