Få mere at vide
3,3’,5-Triiodo-L-thyronine 4’-O-Sulfate, TRC
Artikelnummer. 30006037
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30006037 25 mg -
30007063 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006037 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79539525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 31135-55-4
Molekylær formel C15H12I3NO7S
Molekylvægt (g/mol) 731.04
Synonym Triiodothyronine Sulfate, 3,5-Diiodo-O-[3-iodo-4-(sulfooxy)phenyl]-L-tyrosine, 3-[4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodophenyl]-L-alanine Hydrogen Sulfate, Triiodothyronine Hydrogen Sulfate, Liothyronine Sulfate, T3 Sulfate;
IUPAC navn (2S)-2-amino-3-[3,5-diiodo-4-(3-iodo-4-sulfooxyphenoxy)phenyl]propanoic acid
SMIL C1=CC(=C(C=C1OC2=C(C=C(C=C2I)C[C@@H](C(=O)O)N)I)I)OS(=O)(=O)O
Tekniske data
Kemisk navn eller materiale 3,3',5-Triiodo-L-thyronine 4'-O-Sulfate
Renhedsgrad noter HPLC
InChI formel InChI=1S/C15H12I3NO7S/c16-9-6-8(1-2-13(9)26-27(22,23)24)25-14-10(17)3-7(4-11(14)18)5-12(19)15(20)21/h1-4,6,12H,5,19H2,(H,20,21)(H,22,23,24)/t12-/m0/s1
Mængde 25 mg
Formel vægt 730.7469
Procent renhed >95%