Få mere at vide
3,3’,5-Triiodo-D-thyronine, TRC
Artikelnummer. 30134102
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134102 100 mg
30012373 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134102 Leverandør Toronto Research Chemicals Leverandørnr. TRCT795375100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5714-08-9
Molekylær formel C15 H12 I3 N O4
Molekylvægt (g/mol) 650.97
Synonym D-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]alanine, (+)-Triiodothyronine, 3,5,3'-D-Triiodothyronine, 3,5,3'-Triiodo-D-thyronine, D-3,5,3'-Triiodothyronine, D-Triiodothyronin, D-Triiodothyronine, DT 3, Detrothyronine, Dextrotriiodothyronine, NSC 46046, (R)-2-Amino-3-(4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl)propanoic Acid
IUPAC navn (2R)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoic acid
SMIL N[C@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C(=O)O
Tekniske data
Kemisk navn eller materiale 3,3',5-Triiodo-D-thyronine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C15H12I3NO4/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22/h1-4,6,12,20H,5,19H2,(H,21,22)/t12-/m1/s1
Mængde 100 mg
Formel vægt 650.79
Procent renhed >95%