missing translation for 'onlineSavingsMsg'
Learn More

3-(3,5-Difluorophenyl)-1H-pyrazole-4-carboxaldehyde, 98%, Thermo Scientific™

Catalog Number 15447555
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
875664-59-8
C10H6F2N2O
208.17
MFCD06797905
WYPNQEHRMYDLFU-UHFFFAOYSA-N
3-3,5-difluorophenyl-1h-pyrazole-4-carbaldehyde, 3-3,5-difluorophenyl-1h-pyrazole-4-carboxaldehyde, 5-3,5-difluorophenyl-1h-pyrazole-4-carbaldehyde, 3-3,5-difluorophenyl-2h-pyrazole-4-carbaldehyde, 3-3,5-difluorophenyl pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,3-3,5-difluorophenyl
2783157
5-(3,5-difluorophenyl)-1H-pyrazole-4-carbaldehyde
FC1=CC(=CC(F)=C1)C1=C(C=O)C=NN1
This item is not returnable. View return policy
875664-59-8
C10H6F2N2O
208.17
MFCD06797905
WYPNQEHRMYDLFU-UHFFFAOYSA-N
3-3,5-difluorophenyl-1h-pyrazole-4-carbaldehyde, 3-3,5-difluorophenyl-1h-pyrazole-4-carboxaldehyde, 5-3,5-difluorophenyl-1h-pyrazole-4-carbaldehyde, 3-3,5-difluorophenyl-2h-pyrazole-4-carbaldehyde, 3-3,5-difluorophenyl pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,3-3,5-difluorophenyl
2783157
5-(3,5-difluorophenyl)-1H-pyrazole-4-carbaldehyde
FC1=CC(=CC(F)=C1)C1=C(C=O)C=NN1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 178°C to 181°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 208.16
Percent Purity 98%
Chemical Name or Material 3-(3,5-Difluorophenyl)-1H-pyrazole-4-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'