missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetrakis(trifluoromethyl)benzhydrol, 97%, Thermo Scientific™

Catalog Number 11498927
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
87901-76-6
C17H8F12O
456.23
MFCD00042475
DUCQGUCBWHWEBT-UHFFFAOYSA-N
3,3',5,5'-tetrakis trifluoromethyl benzhydrol, bis 3,5-bis trifluoromethyl phenyl methanol, maybridge1_002893, di 3,5-di trifluoromethyl phenyl methanol, 3,3,5,5-tetrakis trifluoromethyl benzhydrol, benzenemethanol, a-3,5-bis trifluoromethyl phenyl-3,5-bis trifluoromethyl
2776818
bis[3,5-bis(trifluoromethyl)phenyl]methanol
OC(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
This item is not returnable. View return policy
87901-76-6
C17H8F12O
456.23
MFCD00042475
DUCQGUCBWHWEBT-UHFFFAOYSA-N
3,3',5,5'-tetrakis trifluoromethyl benzhydrol, bis 3,5-bis trifluoromethyl phenyl methanol, maybridge1_002893, di 3,5-di trifluoromethyl phenyl methanol, 3,3,5,5-tetrakis trifluoromethyl benzhydrol, benzenemethanol, a-3,5-bis trifluoromethyl phenyl-3,5-bis trifluoromethyl
2776818
bis[3,5-bis(trifluoromethyl)phenyl]methanol
OC(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 147°C to 149°C
Quantity 1 g
Formula Weight 456.23
Percent Purity 97%
Chemical Name or Material 3,3',5,5'-Tetrakis(trifluoromethyl)benzhydrol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'