Få mere at vide
3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Pentadecafluoro-2-nonanol (>90%), TRC
Artikelnummer. 30417014
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30417014 50 mg
1 options
Artikelnummer. 30417014 Leverandør Toronto Research Chemicals Leverandørnr. TRCP27043050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Pentadecafluoro-2-nonanol
CAS 24015-83-6
Molekylær formel C9 H5 F15 O
Mængde 50 mg
Synonym 1-Perfluoroheptyl Ethanol, 2-Nonanol','3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-pentadecafluoro- (8CI','9CI','ACI), 3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Pentadecafluoro-2-nonanol (ACI), 7:2 SFTOH, 1H,1H,1H,2H-Perfluoro-2-nonanol, 1-Perfluoroheptyl Ethanol, 7:2sFTOH
InChI formel InChI=1S/C9H5F15O/c1-2(25)3(10,11)4(12,13)5(14,15)6(16,17)7(18,19)8(20,21)9(22,23)24/h2,25H,1H3
SMIL CC(O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC navn 3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-pentadecafluorononan-2-ol
Molekylvægt (g/mol) 414.11
Formel vægt 414.0101