Få mere at vide
3’-[3-(2,3-Dihydro-2-oxo-1H-benzimidazol-1-yl)propyl] Domperidone(Domperidone Impurity E) (>90%), TRC
Artikelnummer. 30005966
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005966 10 mg
30004215 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30005966 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44946510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346602-50-3
Molekylær formel C32 H34 Cl N7 O3
Molekylvægt (g/mol) 600.11
Synonym 2H-Benzimidazol-2-one, 1-[3-[4-(5-chloro-2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)-1-piperidinyl]propyl]-3-[3-(2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)propyl]-1,3-dihydro-, 1-[3-[4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)piperidin-1-yl]propyl]-3-[3-(2-oxo-2,3-dihydrobenzimidazol-1-yl)-propyl]-1,3-dihydro-2H-benzimidazol-2-one, Domperidone EP Impurity E
IUPAC navn 1-[3-[4-(5-chloro-2-oxo-3H-benzimidazol-1-yl)piperidin-1-yl]propyl]-3-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]benzimidazol-2-one
SMIL Clc1ccc2N(C3CCN(CCCN4C(=O)N(CCCN5C(=O)Nc6ccccc56)c7ccccc47)CC3)C(=O)Nc2c1
Tekniske data
Kemisk navn eller materiale 1-[3-[4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)piperidin-1-yl]propyl]-3-[3-(2-oxo-2,3-dihydrobenzimidazol-1-yl)-propyl]-1,3-dihydro-2H-benzimidazol-2-one
Mængde 10 mg
InChI formel InChI=1S/C32H34ClN7O3/c33-22-11-12-27-25(21-22)35-31(42)40(27)23-13-19-36(20-14-23)15-5-17-38-28-9-3-4-10-29(28)39(32(38)43)18-6-16-37-26-8-2-1-7-24(26)34-30(37)41/h1-4,7-12,21,23H,5-6,13-20H2,(H,34,41)(H,35,42)
Formel vægt 599.2412