Få mere at vide
3,3,17,17-Bis(ethylenedioxy)-19-hydroxyandrost-5-ene-19-d2, TRC
Artikelnummer. 30334371
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30334371 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334371 Leverandør Toronto Research Chemicals Leverandørnr. TRCB4338025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,3,17,17-Bis(ethylenedioxy)-19-hydroxyandrost-4-ene-19-d2
CAS 5696-47-9
Molekylær formel C23H32D2O5
Mængde 5 mg
InChI formel InChI=1S/C23H34O5/c1-20-6-4-19-17(18(20)5-7-23(20)27-12-13-28-23)3-2-16-14-22(25-10-11-26-22)9-8-21(16,19)15-24/h2,17-19,24H,3-15H2,1H3/t17-,18-,19-,20-,21+/m0/s1/i15D2
SMIL [2H]C([2H])(O)[C@]12CCC3(CC1=CC[C@H]4[C@@H]5CCC6(OCCO6)[C@@]5(C)CC[C@H]24)OCCO3
Molekylvægt (g/mol) 392.53
Formel vægt 392.2532