Få mere at vide
3-[[(2S,3R)-2-[(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholino]methyl]-1H-1,2,4-triazol-5(4H)-one ((S,R,S)-Aprepitant), Mikromol™
Artikelnummer. 30049297
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049297 25 mg
30052307 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049297 Leverandør Mikromol Leverandørnr. MM3251.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-[[(2S,3R)-2-[(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholino]methyl]-1H-1,2,4-triazol-5(4H)-one ((S,R,S)-Aprepitant)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (S,R,S)-Aprepitant
CAS 172822-29-6
Analyt- eller komponentnavne (S,R,S)-Aprepitant
Forsendelsestilstand Room Temperature
Molekylær formel C23 H21 F7 N4 O3
Certificeringer/compliance ISO 17025
Synonym 3H-1,2,4-Triazol-3-one, 5-[[(2S,3R)-2-[(1S)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-, ent-Aprepitant, Aprepitant Related Compound B (USP), 3-[[(2S,3R)-2-[(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholino]methyl]-1H-1,2,4-triazol-5(4H)-one, 3H-1,2,4-Triazol-3-one, 5-[[2-[1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-, [2S-[2alpha(R*),3alpha]]-, Aprepitant Related Compound B
InChI formel InChI=1S/C23H21F7N4O3/c1-12(14-8-15(22(25,26)27)10-16(9-14)23(28,29)30)37-20-19(13-2-4-17(24)5-3-13)34(6-7-36-20)11-18-31-21(35)33-32-18/h2-5,8-10,12,19-20H,6-7,11H2,1H3,(H2,31,32,33,35)/t12-,19+,20-/m0/s1
SMIL C[C@H](O[C@@H]1OCCN(CC2=NNC(=O)N2)[C@@H]1c3ccc(F)cc3)c4cc(cc(c4)C(F)(F)F)C(F)(F)F
IUPAC navn 3-[[(2S,3R)-2-[(1S)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholin-4-yl]methyl]-1,4-dihydro-1,2,4-triazol-5-one
Molekylvægt (g/mol) 534.43
Mængde 25 mg
Formel vægt 534.1502 g/mol