missing translation for 'onlineSavingsMsg'
Learn More

3-[[(2S)-1-Methyl-2-pyrrolidinyl]methyl]-5-[(1E)-2-(phenylsulfonyl)ethenyl]-1H-indole, TRC

Product Code. 30483103
Change view
Click to view available options
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30483103 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483103 Supplier Toronto Research Chemicals Supplier No. TRCM3291355MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-(((2S)-1-Methyl-2-pyrrolidinyl)methyl)-5-((1E)-2-(phenylsulfonyl)ethenyl)-1H-indole
CAS 362595-66-2
Molekylær formel C10 H14 N3 O . I
Mængde 5 mg
InChI formel InChI=1S/C10H14N3O.HI/c1-12-6-2-4-9(8-12)10-5-3-7-13(10)11-14;/h2,4,6,8,10H,3,5,7H2,1H3;1H/q+1;/p-1
SMIL [I-].C[n+]1cccc(c1)C2CCCN2N=O
Molekylvægt (g/mol) 319.142
Formel vægt 319.018
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.