Få mere at vide
3-[2-(Trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™
Artikelnummer. 15474537
Skift visning
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15474537 5 g 5g
15464537 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 15474537 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H31876.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 94022-99-8
Molekylær formel C10H9F3O2
Molekylvægt (g/mol) 218.175
MDL nummer MFCD06823965
InChI nøgle YTDVNFDGHHHPEE-UHFFFAOYSA-N
Synonym 3-2-trifluoromethyl phenyl propanoic acid, 3-2-trifluoromethyl phenyl propionic acid, 2-trifluoromethyl hydrocinnamic acid, 2-trifluoromethyl benzenepropanoic acid, benzenepropanoic acid, 2-trifluoromethyl, 3-o-alpha,alpha,alpha-trifluorotolyl propionic acid, acmc-20a31j, 3-2-trifluoromethyl phenyl p, 2-2-carboxyethyl benzotrifluoride
PubChem CID 15284809
ChEBI CHEBI:60707
IUPAC navn 3-[2-(trifluoromethyl)phenyl]propanoic acid
SMIL C1=CC=C(C(=C1)CCC(=O)O)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 3-[2-(Trifluoromethyl)phenyl]propionic acid
Smeltepunkt 79°C to 86°C
Mængde 5 g
Formel vægt 218.17
Procent renhed 97%