missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Thienyl)-2(1H)-quinoxalinone, 97%, Thermo Scientific™

Catalog Number p-7038954
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
64532-10-1
C12H8N2OS
228.269
MFCD00663403
PWIKCZVHLJFOFW-UHFFFAOYSA-N
723698
3-thiophen-2-yl-1H-quinoxalin-2-one
C1=CC=C2C(=C1)NC(=O)C(=N2)C3=CC=CS3
This item is not returnable. View return policy
64532-10-1
C12H8N2OS
228.269
MFCD00663403
PWIKCZVHLJFOFW-UHFFFAOYSA-N
723698
3-thiophen-2-yl-1H-quinoxalin-2-one
C1=CC=C2C(=C1)NC(=O)C(=N2)C3=CC=CS3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Synonym 2 1h-quinoxalinone, 3-2-thienyl, 3-2-thienyl-2 1h-quinoxalinone, 3-2-thienyl-1,2-dihydroquinoxalin-2-one, 3-thiophen-2-yl-1h-quinoxalin-2-one, maybridge3_004430, 3 2-thienyl quinoxalin-2 1h-one, 3 2-thienyl quinoxalin-2 1 h-one, 3-2-thienyl-1h-quinoxalin-2-one, 3-2-thienyl quinoxalin-2 1h-one, 2 1h-quinoxalinone,3-2-thienyl
Formula Weight 228.27
Percent Purity 97%
Chemical Name or Material 3-(2-Thienyl)-2(1H)-quinoxalinone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'