Få mere at vide
3-(2-Hydroxyethyl)benzoic Acid, TRC
Artikelnummer. 30135174
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135174 1 g
30135773 500 mg
17911745 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135174 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9419981G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 320730-08-3
Molekylær formel C9 H10 O3
Molekylvægt (g/mol) 166.17
Synonym 3-(2-Hydroxyethyl)benzoic Acid, Ketoprofen EP Impurity G
IUPAC navn 3-(2-hydroxyethyl)benzoic acid
SMIL OCCc1cccc(c1)C(=O)O
Tekniske data
Kemisk navn eller materiale 3-(2-Hydroxyethyl)benzoic Acid
Mængde 1 g
InChI formel InChI=1S/C9H10O3/c10-5-4-7-2-1-3-8(6-7)9(11)12/h1-3,6,10H,4-5H2,(H,11,12)
Formel vægt 166.063