missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)propanoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-2698989
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
935-13-7
C7H8O3
140.14
XLTJXJJMUFDQEZ-UHFFFAOYSA-N
3-furan-2-yl propanoic acid, 3-2-furyl propanoic acid, 2-furanpropanoic acid, 2-furanpropionic acid, 3-2-furyl propionic acid, 3-furan-2-yl-propionic acid, furan-2-propionic acid, 3-2-furyl propoinic acid, furfuryl-essigsaure, furfurylacetic acid
70286
3-(furan-2-yl)propanoic acid
C1=COC(=C1)CCC(=O)O
This item is not returnable. View return policy
935-13-7
C7H8O3
140.14
XLTJXJJMUFDQEZ-UHFFFAOYSA-N
3-furan-2-yl propanoic acid, 3-2-furyl propanoic acid, 2-furanpropanoic acid, 2-furanpropionic acid, 3-2-furyl propionic acid, 3-furan-2-yl-propionic acid, furan-2-propionic acid, 3-2-furyl propoinic acid, furfuryl-essigsaure, furfurylacetic acid
70286
3-(furan-2-yl)propanoic acid
C1=COC(=C1)CCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 56°C to 59°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol,acetone,ethylether,chlorofor
Formula Weight 140.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Furyl)propanoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'