missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Fluorophenyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog Number p-7032593
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
1643-26-1
C9H9FO2
168.167
MFCD01310820
GUZLQEOSDXLCKX-UHFFFAOYSA-N
2063866
3-(2-fluorophenyl)propanoic acid
C1=CC=C(C(=C1)CCC(=O)O)F
This item is not returnable. View return policy
1643-26-1
C9H9FO2
168.167
MFCD01310820
GUZLQEOSDXLCKX-UHFFFAOYSA-N
2063866
3-(2-fluorophenyl)propanoic acid
C1=CC=C(C(=C1)CCC(=O)O)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 75°C to 79°C
Quantity 1 g
Synonym 3-2-fluorophenyl propionic acid, 3-2-fluorophenyl propanoic acid, benzenepropanoic acid, 2-fluoro, 3-2-fluorophenyl proprionic acid, 3-2-fluoro-phenyl-propionic acid, 2-fluorohydrocinnamic acid, acmc-1bz7v, 2'-fluorocinnamide, 2-fluoro-benzenepropanoic acid, benzenepropanoic acid,2-fluoro
Formula Weight 168.17
Percent Purity 96%
Chemical Name or Material 3-(2-Fluorophenyl)propionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'