missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2-Fluorophenyl)-1-methyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63070.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15469548

  • 8022.97 DKK / 5g


Estimated Shipment date
from Supplier 15-01-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27665229
3-(3-Fluorophenyl)-1-methyl-2-thioxo-4-imidazolidinone

Specifications

3-(2-Fluorophenyl)-1-methyl-2-thiohydantoin
MFCD27665229
224.25
95%
5g
3-(3-Fluorophenyl)-1-methyl-2-thioxo-4-imidazolidinone
Solid
Videos
SDS