Få mere at vide
3-[[[2-[(Diaminomethylene)amino]-4-thiazolyl]methyl]thio]propanimidamide Hydrochloride, TRC
Artikelnummer. 30362684
Skift visning
Mængde:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362684 250 mg
30353978 10 mg
30362449 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30362684 Leverandør Toronto Research Chemicals Leverandørnr. TRCD417670250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76833-47-1
Molekylær formel C8 H14 N6 S2 . Cl H
Molekylvægt (g/mol) 294.83
Synonym Propanimidamide, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-, hydrochloride (1:1), Propanimidamide, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-, monohydrochloride (9CI), 3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]-methyl]sulfanyl]propanimidamide Hydrochloride, Famotidine Imp. A (EP) as Hydrochloride
IUPAC navn 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]propanimidamide;hydrochloride
SMIL Cl.NC(=Nc1nc(CSCCC(=N)N)cs1)N
Tekniske data
Kemisk navn eller materiale 3-[([2-[(Diaminomethylene)amino]thiazol-4-yl]-methyl)sulfanyl]propanimidamideHydrochloride
Mængde 250 mg
InChI formel InChI=1S/C8H14N6S2.ClH/c9-6(10)1-2-15-3-5-4-16-8(13-5)14-7(11)12;/h4H,1-3H2,(H3,9,10)(H4,11,12,13,14);1H
Formel vægt 294.0488