missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2-Chlorophenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56353.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402646

  • 4993.71 DKK / 1g


Estimated Shipment date
from Supplier 19-11-2018
Add to basket

Specifications

Chemical Identifiers

2096353-57-8
293.49
YQDLSLAWMCWVTD-UHFFFAOYSA-N
73996232
C13H10BClFNO3
MFCD22682866
3-2-chlorophenylcarbamoyl-2-fluorobenzeneboronic acid

Specifications

3-(2-Chlorophenylcarbamoyl)-2-fluorobenzeneboronic acid
2096353-57-8
MFCD22682866
YQDLSLAWMCWVTD-UHFFFAOYSA-N
73996232
Solid
1g
C13H10BClFNO3
3-2-chlorophenylcarbamoyl-2-fluorobenzeneboronic acid
293.49
293.48
97%
Videos
SDS