Få mere at vide
3-((2-Chloroethyl)amino)propyl Dihydrogen Phosphate, TRC
Artikelnummer. 30362559
Skift visning
Mængde:
10 mg
20 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362559 10 mg
30355841 50 mg
30350507 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30362559 Leverandør Toronto Research Chemicals Leverandørnr. TRCC29333010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22608-58-8
Molekylær formel C5 H13 Cl N O4 P
Molekylvægt (g/mol) 217.59
Synonym 1-Propanol, 3-[(2-chloroethyl)amino]-, dihydrogen phosphate (ester) (8CI,9CI), Ifosfamide Imp. A
IUPAC navn 3-(2-chloroethylamino)propyl dihydrogen phosphate
SMIL OP(=O)(O)OCCCNCCCl
Tekniske data
Kemisk navn eller materiale 3-[(2-Chloroethyl)amino]propyl Dihydrogen Phosphate
Mængde 10 mg
InChI formel InChI=1S/C5H13ClNO4P/c6-2-4-7-3-1-5-11-12(8,9)10/h7H,1-5H2,(H2,8,9,10)
Formel vægt 217.0271