Få mere at vide
3-(2-Chlorobenzoyl)-4-hydroxy-2H-1-benzopyran-2-one, TRC
Artikelnummer. 30363899
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363899 250 mg
30349366 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30363899 Leverandør Toronto Research Chemicals Leverandørnr. TRCC463415250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-(2-Chlorobenzoyl)-4-hydroxy-2H-1-benzopyran-2-one
CAS 181938-56-7
Molekylær formel C16H9ClO4
Mængde 250 mg
Synonym NSC 133803
InChI formel InChI=1S/C16H9ClO4/c17-11-7-3-1-5-9(11)14(18)13-15(19)10-6-2-4-8-12(10)21-16(13)20/h1-8,19H
SMIL OC1=C(C(=O)c2ccccc2Cl)C(=O)Oc3ccccc13
IUPAC navn 3-(2-chlorobenzoyl)-4-hydroxychromen-2-one
Molekylvægt (g/mol) 300.69
Formel vægt 300.0189