Learn More
3-(2-Benzothiazolyl)aniline, 96%, Thermo Scientific Chemicals
Catalog Number p-7031864
Click to view available options
Quantity:
250 mg
1 g
41230-21-1
C13H10N2S
226.297
MFCD00579111
MKMRZZJJDIBAJF-UHFFFAOYSA-N
3-benzothiazol-2-yl-phenylamine, 3-1,3-benzothiazol-2-yl aniline, 3-benzothiazol-2-ylphenylamine, benzenamine,3-2-benzothiazolyl, cambridge id 5250055, 3-2-benzothiazolyl aniline, 3-benzothiazol-2-yl aniline, 3-benzo d thiazol-2-yl aniline, 3-benzothiazol-2-yl benzenamine, 3-1,3-benzothiazol-2-yl phenyl amine
720999
3-(1,3-benzothiazol-2-yl)aniline
C1=CC=C2C(=C1)N=C(S2)C3=CC(=CC=C3)N
41230-21-1
C13H10N2S
226.297
MFCD00579111
MKMRZZJJDIBAJF-UHFFFAOYSA-N
3-benzothiazol-2-yl-phenylamine, 3-1,3-benzothiazol-2-yl aniline, 3-benzothiazol-2-ylphenylamine, benzenamine,3-2-benzothiazolyl, cambridge id 5250055, 3-2-benzothiazolyl aniline, 3-benzothiazol-2-yl aniline, 3-benzo d thiazol-2-yl aniline, 3-benzothiazol-2-yl benzenamine, 3-1,3-benzothiazol-2-yl phenyl amine
720999
3-(1,3-benzothiazol-2-yl)aniline
C1=CC=C2C(=C1)N=C(S2)C3=CC(=CC=C3)N
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 135°C to 137°C
Quantity 250 mg
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 226.3
Percent Purity 96%
Chemical Name or Material 3-(2-Benzothiazolyl)aniline