missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethyl)-5-methylindole hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7039912
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1010-95-3
C11H15ClN2
210.71
MFCD00012683
RBHDFGBPJGEYCK-UHFFFAOYSA-N
5-methyltryptamine hydrochloride, 2-5-methyl-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methylindole hydrochloride, 5-methyl-1h-indole-3-ethylamine monohydrochloride, 5-methyltryptamine hcl, 1h-indole-3-ethanamine, 5-methyl-, monohydrochloride, 2-5-methylindol-3-yl ethylamine, chloride, 5-methyltryptaminehydrochloride, 5-methyl-1h-indole-3-ethylamine hcl, pubchem24287
6917145
hydrogen 2-(5-methyl-1H-indol-3-yl)ethan-1-amine chloride
[H+].[Cl-].CC1=CC=C2NC=C(CCN)C2=C1
This item is not returnable. View return policy
1010-95-3
C11H15ClN2
210.71
MFCD00012683
RBHDFGBPJGEYCK-UHFFFAOYSA-N
5-methyltryptamine hydrochloride, 2-5-methyl-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methylindole hydrochloride, 5-methyl-1h-indole-3-ethylamine monohydrochloride, 5-methyltryptamine hcl, 1h-indole-3-ethanamine, 5-methyl-, monohydrochloride, 2-5-methylindol-3-yl ethylamine, chloride, 5-methyltryptaminehydrochloride, 5-methyl-1h-indole-3-ethylamine hcl, pubchem24287
6917145
hydrogen 2-(5-methyl-1H-indol-3-yl)ethan-1-amine chloride
[H+].[Cl-].CC1=CC=C2NC=C(CCN)C2=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 290°C to 292°C (decomposition)
Quantity 5 g
Beilstein 3707780
Sensitivity Hygroscopic
Formula Weight 210.71
Percent Purity 97%
Chemical Name or Material 3-(2-Aminoethyl)-5-methylindole hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'