missing translation for 'onlineSavingsMsg'
Learn More

3-[2-[4-[(Z)-(2,4-Difluorophenyl)(hydroxyimino)methyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, Mikromol™

Product Code. 30050621
Change view
Click to view available options
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30050621 100 mg
30049984 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050621 Supplier Mikromol Supplier No. MM0491.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-[2-[4-[(Z)-(2,4-Difluorophenyl)(hydroxyimino)methyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-[2-[4-[(Z)-(2,4-Difluorophenyl)(hydroxyimino)methyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
CAS 132961-05-8
Analyt- eller komponentnavne 3-[2-[4-[(Z)-(2,4-Difluorophenyl)(hydroxyimino)methyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
Forsendelsestilstand Room Temperature
Molekylær formel C23 H28 F2 N4 O2
Certificeringer/compliance ISO 17025
Synonym 3-[2-[4-[(Z)-(2,4-Difluorophenyl)(hydroxyimino)methyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, Risperidone Imp. B (EP), Risperidone Impurity B
InChI formel InChI=1S/C23H28F2N4O2/c1-15-18(23(30)29-10-3-2-4-21(29)26-15)9-13-28-11-7-16(8-12-28)22(27-31)19-6-5-17(24)14-20(19)25/h5-6,14,16,31H,2-4,7-13H2,1H3/b27-22-
SMIL CC1=C(CCN2CCC(CC2)\C(=N\O)\c3ccc(F)cc3F)C(=O)N4CCCCC4=N1
IUPAC navn 3-[2-[4-[(Z)-C-(2,4-difluorophenyl)-N-hydroxycarbonimidoyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
Molekylvægt (g/mol) 430.49
Mængde 100 mg
Formel vægt 430.218 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.