missing translation for 'onlineSavingsMsg'
Få mere at vide

3-[2,4-Bis(trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™

Artikelnummer. p-7031501
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 g
Pakningsstørrelse:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15458135 250 mg 250mg
15448135 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15458135 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H32595.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as intermediates. Also as laboratory reagents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1092460-63-3
Molekylær formel C11H8F6O2
Molekylvægt (g/mol) 286.17
MDL nummer MFCD11519312
InChI nøgle POPIIFPERMLVHR-UHFFFAOYSA-N
Synonym 3-2,4-bis trifluoromethyl phenyl propionic acid, 3-2,4-bis trifluoromethyl phenyl propanoic acid, 2,4-bis trifluoromethyl phenyl propanoic acid, 3-2,4-bis trifluoromethyl phenyl propionic acid, jrd
PubChem CID 54772349
IUPAC navn 3-[2,4-bis(trifluoromethyl)phenyl]propanoic acid
SMIL OC(=O)CCC1=C(C=C(C=C1)C(F)(F)F)C(F)(F)F

Tekniske data

Kemisk navn eller materiale 3-[2,4-Bis(trifluoromethyl)phenyl]propionic acid
Smeltepunkt 74°C to 79°C
Mængde 250 mg
Formel vægt 286.17
Procent renhed 97%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.