Få mere at vide
3-[2-[4-(2,4-Difluorobenzoyl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, Mikromol™
Artikelnummer. 30052541
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052541 25 mg
17931746 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052541 Leverandør Mikromol Leverandørnr. MM0491.100025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-[2-[4-(2,4-Difluorobenzoyl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-[2-[4-(2,4-Difluorobenzoyl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
CAS 158697-67-7
Analyt- eller komponentnavne 3-[2-[4-(2,4-Difluorobenzoyl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
Forsendelsestilstand Room Temperature
Molekylær formel C23 H27 F2 N3 O2
Certificeringer/compliance ISO 17025
Synonym Risperidone Imp. H (EP), 3-[2-[4-(2,4-Difluorobenzoyl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, Risperidone Impurity H
InChI formel InChI=1S/C23H27F2N3O2/c1-15-18(23(30)28-10-3-2-4-21(28)26-15)9-13-27-11-7-16(8-12-27)22(29)19-6-5-17(24)14-20(19)25/h5-6,14,16H,2-4,7-13H2,1H3
SMIL CC1=C(CCN2CCC(CC2)C(=O)c3ccc(F)cc3F)C(=O)N4CCCCC4=N1
IUPAC navn 3-[2-[4-(2,4-difluorobenzoyl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
Molekylvægt (g/mol) 415.48
Mængde 25 mg
Formel vægt 415.2071 g/mol