Få mere at vide
3-[2-(2-Chloroethylamino)ethylamino]propyl Dihydrogen Phosphate Dihydrochloride, Mikromol™
Beskrivelse
3-[2-(2-Chloroethylamino)ethylamino]propyl Dihydrogen Phosphate Dihydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-[2-(2-chloroethylamino)ethylamino]propyl dihydrogen phosphate dihydrochloride |
| CAS | 158401-51-5 |
| Analyt- eller komponentnavne | 3-[2-(2-chloroethylamino)ethylamino]propyl dihydrogen phosphate dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C7 H18 Cl N2 O4 P . 2 Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 1-Propanol, 3-[[2-[(2-chloroethyl)amino]ethyl]amino]-, dihydrogen phosphate (ester), dihydrochloride (9CI), Cyclophosphamide Related Compound D as Dihydrochloride, Cyclophosphamide Impurity E |
| InChI formel | InChI=1S/C7H18ClN2O4P.2ClH/c8-2-4-10-6-5-9-3-1-7-14-15(11,12)13;;/h9-10H,1-7H2,(H2,11,12,13);2*1H |
| SMIL | Cl.Cl.OP(=O)(O)OCCCNCCNCCCl |
| IUPAC navn | 3-[2-(2-chloroethylamino)ethylamino]propyl dihydrogen phosphate;dihydrochloride |
| Vis mere |