Få mere at vide
3-[(1S)-1-(Methylamino)ethyl]phenyl N-Ethyl-N-methylcarbamate, Mikromol
Artikelnummer. 30458658
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30458658 100 mg
1 options
Artikelnummer. 30458658 Leverandør Mikromol Leverandørnr. MM1497.110100
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-[(1S)-1-(Methylamino)ethyl]phenyl N-Ethyl-N-methylcarbamate, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-[(1S)-1-(Methylamino)ethyl]phenyl N-Ethyl-N-methylcarbamate
CAS 923035-05-6
Analyt- eller komponentnavne 3-[(1S)-1-(Methylamino)ethyl]phenyl N-Ethyl-N-methylcarbamate
Forsendelsestilstand Room Temperature
Molekylær formel C13 H20 N2 O2
Certificeringer/compliance ISO 9001
Synonym Namino-Desmethyl Rivastigmine, Carbamic acid, N-ethyl-N-methyl-, 3-[(1S)-1-(methylamino)ethyl]phenyl ester, Rivastigmine Tartrate Imp E (EP)
InChI formel InChI=1S/C13H20N2O2/c1-5-15(4)13(16)17-12-8-6-7-11(9-12)10(2)14-3/h6-10,14H,5H2,1-4H3/t10-/m0/s1
Løsningstype Neat
SMIL CCN(C)C(=O)Oc1cccc(c1)[C@H](C)NC
IUPAC navn [3-[(1S)-1-(methylamino)ethyl]phenyl] N-ethyl-N-methylcarbamate
Molekylvægt (g/mol) 236.31
Mængde 100 mg
Formel vægt 236.152 g/mol