Få mere at vide
3-[(1RS)-5-Bromo-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-1-yl]-N,N-dimethylpropan-1-amine Hydrogen Oxalate, Mikromol™
Beskrivelse
3-[(1RS)-5-Bromo-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-1-yl]-N,N-dimethylpropan-1-amine Hydrogen Oxalate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-[(1R)-5-Bromo-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-1-yl]-N,N-dimethylpropan-1-amine Hydrogen Oxalate |
| CAS | 64372-43-6 |
| Analyt- eller komponentnavne | 3-[(1R)-5-Bromo-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-1-yl]-N,N-dimethylpropan-1-amine Hydrogen Oxalate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H21 Br F N O . C2 H2 O4 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Citalopram Hydrobromide Imp. F (EP), Citalopram Imp. F (EP), Citalopram USP Related Compound H, Citalopram USP RC H, 3-[(1RS)-5-Bromo-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-1-yl]-N,N-dimethylpropan-1-amine Hydrogen Oxalate, Citalopram Hydrobromide Imp. F (EP) as Hydrogen Oxalate, Citalopram Related Compound H as Oxalate, Citalopram Hydrobromide Impurity F as Hydrogen Oxalate, Citalopram Impurity F as Hydrogen Oxalate |
| InChI formel | InChI=1S/C19H21BrFNO.C2H2O4/c1-22(2)11-3-10-19(15-4-7-17(21)8-5-15)18-9-6-16(20)12-14(18)13-23-19;3-1(4)2(5)6/h4-9,12H,3,10-11,13H2,1-2H3;(H,3,4)(H,5,6) |
| SMIL | CN(C)CCCC1(OCc2cc(Br)ccc12)c3ccc(F)cc3.OC(=O)C(=O)O |
| IUPAC navn | 3-[5-bromo-1-(4-fluorophenyl)-3H-2-benzofuran-1-yl]-N,N-dimethylpropan-1-amine;oxalic acid |
| Vis mere |