Få mere at vide
3-[(1RS)-1-[[Ethyl(methyl)carbamoyl]amino]ethyl]phenyl N-Ethyl-N-methylcarbamate, Mikromol™
Beskrivelse
3-[(1RS)-1-[[Ethyl(methyl)carbamoyl]amino]ethyl]phenyl N-Ethyl-N-methylcarbamate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-[(1RS)-1-[[Ethyl(methyl)carbamoyl]amino]ethyl]phenyl N-Ethyl-N-methylcarbamate |
| CAS | 1346242-30-5 |
| Analyt- eller komponentnavne | 3-[(1RS)-1-[[Ethyl(methyl)carbamoyl]amino]ethyl]phenyl N-Ethyl-N-methylcarbamate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H25 N3 O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Carbamic acid, N-ethyl-N-methyl-, 3-[1-[[(ethylmethylamino)carbonyl]amino]ethyl]phenyl ester, 3-[1-[[(Ethylmethylamino)carbonyl]amino]ethyl]phenyl N-ethyl-N-methylcarbamate |
| InChI formel | InChI=1S/C16H25N3O3/c1-6-18(4)15(20)17-12(3)13-9-8-10-14(11-13)22-16(21)19(5)7-2/h8-12H,6-7H2,1-5H3,(H,17,20) |
| SMIL | CCN(C)C(=O)NC(C)c1cccc(OC(=O)N(C)CC)c1 |
| IUPAC navn | [3-[1-[[ethyl(methyl)carbamoyl]amino]ethyl]phenyl] N-ethyl-N-methylcarbamate |
| Vis mere |