Få mere at vide
3-((1RS)-1-Aminoethyl)phenyl N-Ethyl-N-methylcarbamate, Mikromol™
Beskrivelse
3-((1RS)-1-Aminoethyl)phenyl N-Ethyl-N-methylcarbamate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-((1RS)-1-Aminoethyl)phenyl N-Ethyl-N-methylcarbamate |
| CAS | 1346242-29-2 |
| Analyt- eller komponentnavne | 3-((1RS)-1-Aminoethyl)phenyl N-Ethyl-N-methylcarbamate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H18 N2 O2 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Carbamic acid, N-ethyl-N-methyl-, 3-(1-aminoethyl)phenyl ester |
| InChI formel | InChI=1S/C12H18N2O2/c1-4-14(3)12(15)16-11-7-5-6-10(8-11)9(2)13/h5-9H,4,13H2,1-3H3 |
| SMIL | CCN(C)C(=O)Oc1cccc(c1)C(C)N |
| IUPAC navn | [3-(1-aminoethyl)phenyl] N-ethyl-N-methylcarbamate |
| Vis mere |