missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-Methyl-1H-pyrazol-5-yl)benzonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 3-1-methyl-1h-pyrazol-5-yl benzonitrile, 3-2-methylpyrazol-3-yl benzonitrile, benzonitrile,3-1-methyl-1h-pyrazol-5-yl, benzonitrile, 3-1-methyl-1h-pyrazol-5-yl, 3-1-methylpyrazol-5-yl benzenecarbonitrile |
Formula Weight | 183.21 |
Percent Purity | 97% |
Chemical Name or Material | 3-(1-Methyl-1H-pyrazol-5-yl)benzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'