Få mere at vide
3-[1-[Di(methyl-d3)amino]ethyl]phenol, TRC
Artikelnummer. 30378791
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30378791 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378791 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47082210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-[1-[Di(methyl-d3)amino]ethyl]phenol
CAS 194930-03-5
Molekylær formel C10H9D6NO
Mængde 10 mg
Synonym Phenol, 3-[1-[di(methyl-d3)amino]ethyl]-, 3-[1-[Di(methyl-d3)amino]ethyl]phenol, 3-[1-(Dimethylamino)ethyl]phenol-D6 (N,N-dimethyl-D6)
InChI formel InChI=1S/C10H15NO/c1-8(11(2)3)9-5-4-6-10(12)7-9/h4-8,12H,1-3H3/i2D3,3D3
SMIL [2H]C([2H])([2H])N(C(C)c1cccc(O)c1)C([2H])([2H])[2H]
Molekylvægt (g/mol) 171.27
Formel vægt 171.153