Learn More
Alfa Aesar™ 3-(1-Cyanoethyl)benzoic acid, 98%
Brand: Alfa Aesar™ H60537.03
Additional Details : CAS Number : 5537-71-3 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
5537-71-3 | |
175.187 | |
IRYIYPWRXROPSX-UHFFFAOYSA-N | |
98370 | |
3-(1-cyanoethyl)benzoic acid |
C10H9NO2 | |
MFCD00002520 | |
3-1-cyanoethyl benzoic acid, m-1-cyanoethyl benzoic acid, benzoic acid, 3-1-cyanoethyl, zlchem 504, acmc-1b01z, 3-1-cyanethyl benzoesaeure, 3-1-cyanoethyl benzoicacid, 3-l-cyanoethyl benzoic acid, 3-1-cyanethyl benzoic acid, meta-1-cyanoethyl benzoic acid | |
CHEBI:85131 | |
CC(C#N)C1=CC=CC(=C1)C(=O)O |
Specifications
3-(1-Cyanoethyl)benzoic acid | |
1g | |
C10H9NO2 | |
3-1-cyanoethyl benzoic acid, m-1-cyanoethyl benzoic acid, benzoic acid, 3-1-cyanoethyl, zlchem 504, acmc-1b01z, 3-1-cyanethyl benzoesaeure, 3-1-cyanoethyl benzoicacid, 3-l-cyanoethyl benzoic acid, 3-1-cyanethyl benzoic acid, meta-1-cyanoethyl benzoic acid | |
CC(C#N)C1=CC=CC(=C1)C(=O)O | |
175.187 | |
CHEBI:85131 | |
98% |
145°C to 148°C | |
5537-71-3 | |
MFCD00002520 | |
IRYIYPWRXROPSX-UHFFFAOYSA-N | |
3-(1-cyanoethyl)benzoic acid | |
98370 | |
175.18 |