Learn More
3-((1-Benzyl-1H-indazol-3-yl)oxy)-N,N-dimethylpropan-1-amine hydrochloride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 132-69-4 |
| Molecular Formula | C19H24ClN3O |
| MDL Number | MFCD00078957 |
| Quantity | 25 g |
| SMILES | CN(C)CCCOC1=NN(CC2=CC=CC=C2)C3=C1C=CC=C3.[H]Cl |
| Molecular Weight (g/mol) | 345.87 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-((1-Benzyl-1H-indazol-3-yl)oxy)-N,N-dimethylpropan-1-amine hydrochloride |