missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1,2,3,6-Tetrahydro-4-pyridyl)indole, 95%

Brand:  Alfa Aesar™ H63821.06

Additional Details : CAS Number : 65347-55-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15490719

  • 4444.94 DKK / 5g


Estimated Shipment
23-10-2020
Add to basket

Specifications

Chemical Identifiers

65347-55-9
198.269
CIRSPTXGPFAXRE-UHFFFAOYSA-N
2761023
C1CNCC=C1C2=CNC3=CC=CC=C32
C13H14N2
MFCD03695471
3-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, chembl27811, 3-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole, 3-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole, 3-4-1,2,5,6-tetrahydropyridyl indole, thpindole, d02mvr, 3-1,2,3,6-tetrahydropyridin-4-yl indole, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indol, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indole
3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole

Specifications

3-(1,2,3,6-Tetrahydro-4-pyridyl)indole
65347-55-9
MFCD03695471
3-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, chembl27811, 3-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole, 3-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole, 3-4-1,2,5,6-tetrahydropyridyl indole, thpindole, d02mvr, 3-1,2,3,6-tetrahydropyridin-4-yl indole, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indol, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indole
C1CNCC=C1C2=CNC3=CC=CC=C32
198.269
198.26
95%
5g
C13H14N2
958099
CIRSPTXGPFAXRE-UHFFFAOYSA-N
3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
2761023
Solid
Videos
SDS Documentos