missing translation for 'onlineSavingsMsg'
Få mere at vide

(2Z)-2-Butenedioic Acid 1-Methyl Ester-d3, TRC

Artikelnummer. 30338161
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30338161 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30338161 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6908625MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (2Z)-2-Butenedioic Acid 1-Methyl Ester-d3
CAS 1266402-66-7
Molekylær formel C5H3D3O4
Mængde 5 mg
Synonym (Z)-Butenedioic Acid Monomethyl Ester-d3, Maleic Acid Methyl Ester-d3, Methyl Hydrogen Maleate-d3, Monomethyl (2Z)-2-Butenedioate-d3, Monomethyl Maleate-d3
InChI formel InChI=1S/C5H6O4/c1-9-5(8)3-2-4(6)7/h2-3H,1H3,(H,6,7)/b3-2-/i1D3
SMIL [2H]C([2H])([2H])OC(=O)\C=C/C(=O)O
IUPAC navn (Z)-4-oxo-4-(trideuteriomethoxy)but-2-enoic acid
Molekylvægt (g/mol) 133.12
Formel vægt 133.0454
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.