Få mere at vide
(2S-trans)-3-Methyl-3-(4-methyl-3-pentenyl)-oxiranemethanol Methanesulfonate, TRC
Artikelnummer. 30476189
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476189 100 mg
30475598 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30476189 Leverandør Toronto Research Chemicals Leverandørnr. TRCM321315100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2S-trans)-3-Methyl-3-(4-methyl-3-pentenyl)-oxiranemethanol Methanesulfonate
CAS 93221-74-0
Molekylær formel C11H20O4S
Mængde 100 mg
Synonym (2S,3S)-3-Methyl-3-(4-methyl-3-penten-1-yl)-2-oxiranemethanol 2-Methanesulfonate
InChI formel InChI=1S/C11H20O4S/c1-9(2)6-5-7-11(3)10(15-11)8-14-16(4,12)13/h6,10H,5,7-8H2,1-4H3/t10-,11-/m0/s1
SMIL CC(=CCC[C@]1(C)O[C@H]1COS(=O)(=O)C)C
IUPAC navn [(2S,3S)-3-methyl-3-(4-methylpent-3-enyl)oxiran-2-yl]methyl methanesulfonate
Molekylvægt (g/mol) 248.34
Formel vægt 248.1082