Få mere at vide
(2S)-Octyl-Alpha-hydroxyglutarate, TRC
Artikelnummer. 17938884
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17938884 10 mg
17978774 1 mg
17937524 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17938884 Leverandør Toronto Research Chemicals Leverandørnr. TRCO28761010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1391194-64-1
Molekylær formel C13 H24 O5
Molekylvægt (g/mol) 260.33
Synonym Pentanedioic acid, 2-hydroxy-, 1-octyl ester, (2S)-, 1-Octyl (2S)-2-hydroxypentanedioate, (S)-4-Hydroxy-5-(octyloxy)-5-oxopentanoic acid, (2S)-2-Hydroxyglutaric acid octyl ester, (2S)-Octyl-2-HG, 2S-Hydroxypentanedioic Acid 1-Octyl Ester
IUPAC navn (4S)-4-hydroxy-5-octoxy-5-oxopentanoic acid
SMIL CCCCCCCCOC(=O)[C@@H](O)CCC(=O)O
Tekniske data
Kemisk navn eller materiale (2S)-Octyl-Alpha-hydroxyglutarate
Mængde 10 mg
InChI formel InChI=1S/C13H24O5/c1-2-3-4-5-6-7-10-18-13(17)11(14)8-9-12(15)16/h11,14H,2-10H2,1H3,(H,15,16)/t11-/m0/s1
Formel vægt 260.1624