Få mere at vide
(2S)-N-Methoxy-N,2-dimethylhexanamide-d3, TRC
Artikelnummer. 30481923
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30481923 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481923 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22612725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2S)-N-Methoxy-N,2-dimethylhexanamide-d3
CAS 1330165-35-9
Molekylær formel C20 H25 F3 O3 S
Mængde 25 mg
Synonym (2S)-N-Methoxy-N-methyl-(2-methyl-d3)-hexanamide, (2S)-N-Methyl-2-methylhexanohydroxamic Acid Methyl Ether-d3
InChI formel InChI=1S/C20H25F3O3S/c1-18-11-4-3-5-13(18)6-7-14-15-8-9-17(19(15,2)12-10-16(14)18)26-27(24,25)20(21,22)23/h3,5-6,9,14-16H,4,7-8,10-12H2,1-2H3/t14-,15+,16+,18-,19-/m0/s1
SMIL C[C@]12CC[C@@H]3[C@@H](CC=C4C=CCC[C@]34C)[C@H]1CC=C2OS(=O)(=O)C(F)(F)F
IUPAC navn [(8R,9R,10R,13S,14R)-10,13-dimethyl-2,7,8,9,11,12,14,15-octahydro-1H-cyclopenta[a]phenanthren-17-yl] trifluoromethanesulfonate
Molekylvægt (g/mol) 402.471
Formel vægt 402.148