Få mere at vide
(2S)-N-(2-Boc-amino-3-phenyl-d5-propyl) Boc-glycine, TRC
Artikelnummer. 30334506
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30334506 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334506 Leverandør Toronto Research Chemicals Leverandørnr. TRCB62111710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2S)-N-(2-Boc-amino-3-phenyl-d5-propyl) Boc-glycine
CAS 1217601-60-9
Molekylær formel C21H27D5N2O6
Mængde 10 mg
Synonym (2S)-N-Boc-N’-Boc-amino-3-(phenyl-d5)propylglycine
InChI formel InChI=1S/C21H32N2O6/c1-20(2,3)28-18(26)22-16(12-15-10-8-7-9-11-15)13-23(14-17(24)25)19(27)29-21(4,5)6/h7-11,16H,12-14H2,1-6H3,(H,22,26)(H,24,25)/t16-/m0/s1/i7D,8D,9D,10D,11D
SMIL [2H]c1c([2H])c([2H])c(C[C@@H](CN(CC(=O)O)C(=O)OC(C)(C)C)NC(=O)OC(C)(C)C)c([2H])c1[2H]
IUPAC navn 2-[(2-methylpropan-2-yl)oxycarbonyl-[(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(2,3,4,5,6-pentadeuteriophenyl)propyl]amino]acetic acid
Molekylvægt (g/mol) 413.52
Formel vægt 413.2574