Få mere at vide
(2S,5S)-5-Benzyl-3,6-dioxo-2-piperazineacetic Acid, TRC
Artikelnummer. 30135171
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135171 250 mg
30133431 50 mg
30003675 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135171 Leverandør Toronto Research Chemicals Leverandørnr. TRCB276305250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5262-10-2
Molekylær formel C13 H14 N2 O4
Molekylvægt (g/mol) 262.26
Synonym 2-(5-Benzyl-3,6-dioxopiperazin-2-yl)acetic Acid, Aspartame Imp. A (EP), Diketopiperazine, 2-Piperazineacetic acid, 3,6-dioxo-5-(phenylmethyl)-, (2S,5S)- (9CI, ACI), (2S,5S)-3,6-Dioxo-5-(phenylmethyl)-2-piperazineacetic acid (ACI), 2-Piperazineacetic acid, 3,6-dioxo-5-(phenylmethyl)-, (2S-cis)- (ZCI), 2-Piperazineacetic acid, 5-benzyl-3,6-dioxo- (7CI, 8CI), Cycloaspartylphenylalanine, (2S,5S)-5-Benzyl-3,6-dioxo-2-piperazineacetic acid, 2-[(2S,5S)-5-Benzyl-3,6-dioxopiperazin-2-yl]acetic acid, (2S-cis)-5-Benzyl-3,6-dioxopiperazine-2-acetic acid
IUPAC navn 2-[(2S,5S)-5-benzyl-3,6-dioxopiperazin-2-yl]acetic acid
SMIL OC(=O)C[C@@H]1NC(=O)[C@H](Cc2ccccc2)NC1=O
Tekniske data
Kemisk navn eller materiale 2-(5-Benzyl-3,6-dioxopiperazin-2-yl)acetic Acid
Analyt- eller komponentnavne 2-(5-Benzyl-3,6-dioxopiperazin-2-yl)acetic Acid
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C13H14N2O4/c16-11(17)7-10-13(19)14-9(12(18)15-10)6-8-4-2-1-3-5-8/h1-5,9-10H,6-7H2,(H,14,19)(H,15,18)(H,16,17)/t9-,10-/m0/s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 250 mg
Formel vægt 262.0954
Procent renhed >95%