Få mere at vide
(2S,5R,6R)-6-[[(2S)-2-Amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (L-Ampicillin), Mikromol™
Artikelnummer. 30050997
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050997 100 mg
30049601 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050997 Leverandør Mikromol Leverandørnr. MM0253.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2S,5R,6R)-6-[[(2S)-2-Amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (L-Ampicillin)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2S,5R,6R)-6-[((2S)-2-Amino-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (L-Ampicillin)
CAS 19379-33-0
Analyt- eller komponentnavne (2S,5R,6R)-6-[((2S)-2-Amino-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (L-Ampicillin)
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C16 H19 N3 O4 S
Certificeringer/compliance ISO 9001
Synonym Ampicillin Sodium Imp. B (EP), Ampicillin Imp. B (EP), (2S,5R,6R)-6-[[(2S)-2-Amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid, L-Ampicillin, Ampicillin Sodium Impurity B, Ampicillin Impurity B
InChI formel InChI=1S/C16H19N3O4S/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23)/t9-,10+,11-,14+/m0/s1
SMIL CC1(C)S[C@@H]2[C@H](NC(=O)[C@@H](N)c3ccccc3)C(=O)N2[C@H]1C(=O)O
IUPAC navn (2S,5R,6R)-6-[[(2S)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molekylvægt (g/mol) 349.4
Mængde 100 mg
Formel vægt 349.1096 g/mol