Få mere at vide
(2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[[2-(4-hydroxyphenoxy)acetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (4-Hydroxyphenoxymethylpenicillin), Mikromol™
Artikelnummer. 30052272
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052272 25 mg
30050636 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052272 Leverandør Mikromol Leverandørnr. MM0026.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[[2-(4-hydroxyphenoxy)acetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (4-Hydroxyphenoxymethylpenicillin)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxyphenoxymethylpenicillin
CAS 20880-67-5
Analyt- eller komponentnavne 4-Hydroxyphenoxymethylpenicillin
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C16 H18 N2 O6 S
Certificeringer/compliance ISO 9001
Synonym Phenoxymethylpenicillin Potassium Imp. D (EP), Phenoxymethylpenicillin Imp. D (EP), (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[[2-(4-hydroxyphenoxy)acetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylicAcid, 4-Hydroxyphenoxymethylpenicillin, Phenoxymethylpenicillin Potassium Impurity D, Phenoxymethylpenicillin Impurity D
InChI formel InChI=1S/C16H18N2O6S/c1-16(2)12(15(22)23)18-13(21)11(14(18)25-16)17-10(20)7-24-9-5-3-8(19)4-6-9/h3-6,11-12,14,19H,7H2,1-2H3,(H,17,20)(H,22,23)/t11-,12+,14-/m1/s1
SMIL CC1(C)S[C@@H]2[C@H](NC(=O)COc3ccc(O)cc3)C(=O)N2[C@H]1C(=O)O
IUPAC navn (2S,5R,6R)-6-[[2-(4-hydroxyphenoxy)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molekylvægt (g/mol) 366.39
Mængde 25 mg
Formel vægt 366.0886 g/mol